Bottles, wide neck, square, with screw cap, Nalgene®
Supplier: Thermo Fisher Scientific
2110-0002 2110-0006 2110-0008 2110-0016 2110-0032 2114-0002 2114-0006 2114-0008 2114-0016 2114-0032
215-7272EA 46.3 EUR
215-7272 215-7273 215-7274 215-7275 215-7276 215-7282 215-7283 215-7284 215-7285 215-7286
Bottles, wide neck, square, with screw cap, Nalgene®
Bottles Plastic Bottles
HDPE, translucent or PPCO, translucent, with PP screw cap.
  • HDPE: Compatibility with most alcohols, bases and acids. PPCO: Very good chemical resistance
  • HDPE: Heat resistant up to +120 °C. PPCO: Heat resistant up to +121 °C, autoclavable
  • Leakproof
Ideal for storing and shipping samples. Wide mouth permits easy filling and cleaning. Square shape saves space.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact,Product Transparency and Disclosure Product Transparency and Disclosure Low Manufacturing Impact,Product Transparency and Disclosure Low Manufacturing Impact,Product Transparency and Disclosure Low Manufacturing Impact,Product Transparency and Disclosure
Environmental certification NAR Certificate of Retirement NAR Certificate of Retirement NAR Certificate of Retirement NAR Certificate of Retirement NAR Certificate of Retirement IRECS; ISO 14001 (Certificate Registration No. EMS 752603), ACT Label ACT Label IRECS; ISO 14001 (Certificate Registration No. EMS 752603), ACT Label IRECS; ISO 14001 (Certificate Registration No. EMS 752603), ACT Label IRECS; ISO 14001 (Certificate Registration No. EMS 752603), ACT Label