Safety wash bottles, right-to-understand, Nalgene®
Supplier: Thermo Fisher Scientific
2429-0501 2429-0502 2429-0503 2429-0504 2429-0505 2429-0506
215-8100EA 570 EUR
215-8100 215-8109 215-8110 215-8119 215-8120 215-8125
Safety wash bottles, right-to-understand, Nalgene®
Bottles Wash Bottles
Narrow, cylindrical FEP bottle with ETFE dispensing cap. New globally harmonised chemical labeling rules have been enacted to protect workers who come into contact with hazardous chemicals.
  • Greater awareness of hazards, resulting in safer use of chemicals in the workplace; detailed safety information at your fingertips
  • 100% GHS information (no mixing of systems): wash bottles are labelled with the same warning system as is used on purchased solvents
  • Chemically inert (low leachable) making it suitable for storing high purity solvent
  • Autoclavable, with stem
More Product Information