Sample containers, with screw cap, universal, WHEATON®
Supplier: DWK Life Sciences
327150 327151 327150PP 327152 327151PP 327152PP 327153 327153PP 327155 327155PP
216-0982EA 148 EUR
216-0982 216-0984 216-0983 216-0986 216-0985 216-0987 216-0988 216-0989 216-0990 216-0991
Sample containers, with screw cap, universal, WHEATON®
Sample Containers General Purpose Containers
PP translucent or PS clear, with PE screw cap.
  • Manufactured from FDA approved virgin polystyrene or polypropylene
  • PS offers good thermal and chemical resistance
  • Supplied sterile - for in vitro use only
  • All materials are non cytotoxic
Straight walled containers ideal for sample collection, storage and transport. Supplied with leakproof, flow seal closure. Available pre-filled with boric acid for urine samples.
Certifications: CE marked in accordance with European Directive 98/79/EC
All containers are leak tested to British Standard BS5213:1975(1989)
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable