Cutting board
Supplier: SciLabware
BWY300
216-1300EA 50 EUR
216-1300
Cutting board
Dissecting Boards
Durable cutting board made of PP.
  • Autoclavable at 121 °C
  • Good chemical resistance
  • Unbreakable in normal use
  • Non absorbent and easy to clean