ThermoSafe® EPS Insulated Shippers
Supplier: Sonoco Thermosafe
11096 11262 50001359 50001915 50002062 50002358 50004243 505710 517230 517225
216-1294EA 624 EUR
216-1294 216-1323 216-1324 216-1325 216-1327 216-1329 216-1331 216-1337 216-1339 SCAT517225
ThermoSafe® EPS Insulated Shippers
Mailers and Shipping Containers
EPS is an effective insulator which protects cargo from external changes in temperature.
  • High-quality EPS base and lid
  • Ideal for the storage and transportation of temperature-sensitive products
  • EPS is an effective insulator which protects cargo from external changes in temperature
  • Use in combination with refrigerants or dry ice to create a bespoke solution for varied application
EPS insulated containers offer the ideal solution for the safe storage and delivery of many refrigerated or frozen materials. They are lightweight yet tough, making them aMore Product Information