VWR®, Solid Euro Containers
Supplier: VWR Collection
216-2390EA 8.3 EUR
216-2390 216-2391 216-2392 216-2393 216-2394 216-2395 216-2396 216-2397 216-2398 216-2399 216-2400 216-2401 216-2402 216-2409 216-2410 216-2416
VWR®, Solid Euro Containers
Boxes Transport Boxes
These plastic stackable Euro containers are ideal for simple storage solutions as well as highly automated systems. Made from strong plastic these containers are ideal for transporting goods and food safely. They are very stable when loaded due to reinforced frame and corner construction.
  • Made of PPC (polypropylene copolymer)
  • Suitable for every industry and for any application
  • Smooth surfaces make them easy to clean
  • Ergonomic handles for effective manual handling
  • Suitable for –20 to +65 °C storage
  • 100% recyclable
The solid Euro containers areMore Product Information