Stoppers, Glass, Conical Ground Joint
Supplier: DWK Life Sciences
216220708
217-0767EA 247 EUR
217-0767
Stoppers, Glass, Conical Ground Joint
Stoppers Glass Stoppers
DURAN® glass stopper with conical ground joint, for reagent bottles and oxygen bottles according to Winkler.
  • Made of borosilicate 3.3 glass