Anti-CD40L (24-31) Mouse Monoclonal Antibody

Supplier: Merck Millipore (Oncogene)
217595-100
217595-100EA 758 EUR
217595-100
Anti-CD40L (24-31) Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: CD40L (24-31)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR