Rack for scintillation vials, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F185120000
218-1290EA 113 EUR
218-1290
Rack for scintillation vials, SP Bel-Art
Racks Scintillation Vial Racks
PP, autoclavable.
  • Two handles for safe and easy transport
  • Two levels for good sample visibility
Rack for 24 scintillation vials (3×8 format).
W×D×H: 298×127×95 mm.