Dropping funnels, cylindrical, without pressure balance
Supplier: JONES FR
WJ590 WJ592 WJ593
221-0075EA 180 EUR
221-0075 221-0076 221-0077
Dropping funnels, cylindrical, without pressure balance
Funnels Dropping Funnels
Borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Cylindrical shape, with standard ground joint socket and stopper. PTFE stopcock and thread guard.
Order Now
SPECIFICATIONS
Environmentally Preferable