VWR®, Funnels with Long Stem
Supplier: VWR Collection
221-0204EA 26.1 EUR
221-0204 221-0205 221-0206 221-0207 221-0208
VWR®, Funnels with Long Stem
Funnels Filtration Funnels Filter Funnels
PTFE, white.
  • Temperature resistance: –200 to +280 °C
  • Totally chemically inert
  • Super smooth non-stick internal finish
Conical opening with long stem.
Order Now
SPECIFICATIONS
Environmentally Preferable