Mercury (II) oxide, yellow ≥99.0% ACS, Fluka™

Supplier: Honeywell Chemicals
Danger

221082-500G 221082-5G
221082-500GEA 366 EUR
221082-500G 221082-5G
Mercury (II) oxide, yellow ≥99.0% ACS, Fluka™
Mercury (II) oxide
Formula: HgO
Melting Pt: 400 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011045
CAS Number: 21908-53-2
EINECS: 244-654-7
UN: 1641
ADR: 6.1,II
Merck Index: 13,05908

Order Now

Specification Test Results

Assay Min. 99.0 %
Glowing After reducing with HCOOH(as SO₄) Max. 0.05 %
Insoluble in HCl Max. 0.03 %
Iron (Fe) Max. 0.003 %
Chloride (Cl) Max. 0.025 %
Sulfate (SO₄) Max. 0.01 %
Total N Max. 0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR