Cadmium acetate dihydrate ≥99.0%, AnalaR NORMAPUR® analytical reagent

Supplier: VWR Chemicals
Warning

Synonyms: Cadmium di(acetate) dihydrate, Acetic acid cadmium salt dihydrate

22189.294EA 281 EUR
22189.294
Cadmium acetate dihydrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Cadmium acetate dihydrate
Formula: (H₃CCOO)₂Cd·2H₂O
MW: 266.53 g/mol
Boiling Pt: 105…107 °C (4 Torr)
Melting Pt: 256
Density: 2.341 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150020
CAS Number: 5743-04-4
EINECS: 208-853-2
UN: 2570
ADR: 6.1 III
Merck Index: 13,01614

Order Now

Specification Test Results

Assay Min. 99.0 %
Cl (Chloride) Max. 20 ppm
SO4 (Sulphate) Max. 50 ppm
Ca (Calcium) Max. 50 ppm
Cu (Copper) Max. 5 ppm
Fe (Iron) Max. 5 ppm
Na (Sodium) Max. 20 ppm
Pb (Lead) Max. 50 ppm
Zn (Zinc) Max. 20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR