Ammonium hydrogen difluoride 95%, Reagent Grade

Supplier: Honeywell Chemicals
Danger

224820-2.5KG 224820-500G
224820-2.5KGEA 423 EUR
224820-2.5KG 224820-500G
Ammonium hydrogen difluoride 95%, Reagent Grade
Ammonium hydrogen difluoride

  • ≥95% for general laboratory use.

Formula: NH₄HF₂
Melting Pt: 126 °C (Sublimation point 1,013 hPa)
Density: 1,500 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012018
CAS Number: 1341-49-7
EINECS: 215-676-4
UN: 1727
ADR: 8,II
Merck Index: 13,00498

Order Now

Specification Test Results

Identity (IR) complying
Assay of NH₄HF₂ 94.0 - 106.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR