Carborundum, TECHNICAL 1.190 mm

Supplier: VWR Chemicals

22490.295EA 268 EUR
22490.295
Carborundum, TECHNICAL 1.190 mm
Carborundum
Formula: SiC
MW: 40.1 g/mol
Boiling Pt: < 2700 °C (1013 hPa)
Melting Pt: 2700 °C
Density: 3.23 g/cm³ (20 °C)
CAS Number: 409-21-2
EINECS: 206-991-8
Merck Index: 12,08636

Order Now

Specification Test Results

Identification Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR