Beeswax, yellow, TECHNICAL

Supplier: VWR Chemicals

22842.268EA 402 EUR
22842.268
Beeswax, yellow, TECHNICAL
Beeswax
Melting Pt: ∼64 °C
Density: 0,95 g/cm³ (20 °C)
MDL Number: MFCD00132754
CAS Number: 8012-89-3
EINECS: 232-383-7

Order Now

Specification Test Results

Melting point 61 - 65 °C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR