Dialysis bag clips, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F182370000
229-0003EA 59.6 EUR
229-0003
Dialysis bag clips, SP Bel-Art
Clamps General Purpose Clamps
These strong clips made from PP are ideal for suspending dialysis bags in large beakers or pitchers up to 0,63 cm (¼") thick.
  • Bags slide easily into the slot and then the holder can be clipped onto the container’s edge
  • Smooth PP surface may be marked with identifying sequence numbers
  • Autoclavable