Dimethyl sulphide ≥99%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: DMS

22949.AK 22949.K2
22949.AKEA 46.8 EUR
22949.AK 22949.K2
Dimethyl sulphide ≥99%
Dimethyl sulphide

Purity: 99+% CAS No.: 75-18-3 Molecular Formula: (CH3)2S Formula Weight: 62.14 Flash Point: -48°(-54°F) Physical Form: Liquid Density: 0.845 Refractive Index: 1.4351 Odour: Unpleasant Melting Point: -83° Boiling Point: 38° Merck Reference: 14,6123 UN No.: 1164 MDL No.: MFCD00008562

Formula: C₂H₆S
MW: 62.14 g/mol
Boiling Pt: 38 °C
Melting Pt: –83 °C
Density: 0.845
Flash Pt: –48 °C (–54 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008562
CAS Number: 75-18-3
EINECS: 200-846-2
UN: 1164
ADR: 3 II
Merck Index: 12,06204

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR