Double spatulas
Supplier: BOCHEM
3100 3102 3103 3105 3107 3104 3108 31005 31015 31025 31035 31055 31045 3101
231-1032EA 4.9 EUR
231-1032 231-1034 231-1035 231-1037 231-1038 231-1036 231-1039 231-0038 231-0039 231-0040 231-0041 231-0043 231-0042 231-1033
Double spatulas
Spatulas
Stainless steel 18/10, anti-magnetic. Multipurpose spatula for a wide range of uses.
  • Round handle, one flat and one rounded end
  • Straight shape
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance