Detectable spoons, SteriPlast®
Supplier: BURKLE
5378-2011 5378-2012
231-0575EA 82 EUR
231-0575 231-0576
Detectable spoons, SteriPlast®
Spoons
These detectable SteriPlast® spoons, made of PS, are produced in a class 7 cleanroom (10,000), individually packaged for disposable use and then sterilised by gamma rays. The detectable spoons are available in two sizes, 2,5 ml (similar to a teaspoon) and 10 ml (similar to a tablespoon).
  • Can be detected by colour scanners, metal detectors or X-rays
  • Individually packaged and sterilised by gamma rays
  • Production, assembly and packaging according to cleanroom class 7
  • Complies with EU food requirements and FDA regulations
These spoons are not only colouredMore Product Information