Sample spoons, PS, double-bagged, long handle, Sterileware®, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
H36940-1010 H36942-1010 H36944-1010 H36946-1010
231-0644EA 188 EUR
231-0644 231-0645 231-0646 231-0647
Sample spoons, PS, double-bagged, long handle, Sterileware®, SP Bel-Art
Spoons
The long handle on these spoons enables users to reach into jars, bottles and other confined areas to minimise the potential of personal contact with sampled material.
  • Ideal for use in multi-zone cleanrooms
  • FDA grade polystyrene safe for food, drug and cosmetic use
  • Gamma irradiated sterile to SAL 10⁻⁶
  • Lot stamped for accurate lot tracing, and shipped with a certificate of processing for sterilisation
Minimises contamination of small sample collection. Ten individually packed scoops are sealed in an outer bag to create multiple layers of protectionMore Product Information