Spoons, POLY
Supplier: BOCHEM
3403 3413 3401 3414 3411 3410 3412 3395 3400 3402 3404
231-1269EA 13.9 EUR
231-1269 231-1270 231-1069 231-0605 231-0604 231-1296 231-1297 231-1261 231-1068 231-1070 231-1071
Spoons, POLY
Spoons
Stainless steel 18/10, anti-magnetic. Multipurpose spoon for a wide range of uses. Special shape for light and easy handling.
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Quick and easy to clean
Order Now
SPECIFICATIONS
Environmentally Preferable