Copper(II) sulphate pentahydrate 99,0-102,0, AnalaR NORMAPUR® Reag. Ph. Eur., ACS zA

Supplier: VWR Chemicals
Danger

Synonyms: Cupric sulphate pentahydrate, Copper sulphate pentahydrate

23174.233EA 48.6 EUR
23174.233 23174.290 23174.360 23174.460
Copper(II) sulphate pentahydrate 99,0-102,0, AnalaR NORMAPUR® Reag. Ph. Eur., ACS zA
Copper (II) sulphate pentahydrate
Formula: CuSO₄·5H₂O
MW: 249.69 g/mol
Density: 2.284 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149681
CAS Number: 7758-99-8
EINECS: 231-847-6
UN: 3077
ADR: 9 III
REACH: 01-2119520566-40
Merck Index: 13,02682

Order Now

Specification Test Results

Assay 99.0 - 102.0 %
Insolubility in water Max. 50 ppm
Total N (Nitrogen) Max. 10 ppm
Cl (Chloride) Max. 5 ppm
Ca (Calcium) Max. 50 ppm
Fe (Iron) Max. 20 ppm
K (Potassium) Max. 10 ppm
Mg (Magnesium) Max. 50 ppm
Na (Sodium) Max. 50 ppm
Ni (Nickel) Max. 50 ppm
Pb (Lead) Max. 50 ppm
Zn (Zinc) Max. 0.03 %
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR