Blades for safety knives, No. 142
Supplier: MARTOR ARGENTAX
142.70
233-2238EA 17.2 EUR
233-2238
Blades for safety knives, No. 142
Razors and Blades
Non corrosive trapezoid blade for use with safety knives.
  • Deep-edged, two-sided ground-edge
  • Double bevel edge-ground
  • Two-times usable blade
Mountable in:
SECUNORM PROFI
SECUNORM IDEAL