Dithiothreitol (DTT, Cleland's reagent), Millipore®

Supplier: Merck Millipore Calbiochem
Warning

233156-1 233156-10
233156-1EA 169 EUR
233156-1 233156-10
Dithiothreitol (DTT, Cleland's reagent), Millipore®
Dithiothreitol (DTT, Cleland's reagent)
Formula: C₄H₁₀O₂S₂
Boiling Pt: ∼125 °C (1013 hPa)
Melting Pt: 42…44 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004877
CAS Number: 3483-12-3
EINECS: 222-468-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5022472c3f3baef75a703791c548fc6504df810f&vid=81529732db015cb66c1f106092309ae5714325b7&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR