Mixing measuring cylinders, tall form, class A, borosilicate glass
Supplier: HIRSCHMANN
2340194 2340160 2340170 2340175 2340180 2340186 2340190 2340193
2340194.EA 227 EUR
2340194. 2340160. 2340170. 2340175. 2340180. 2340186. 2340190. 2340193.
Mixing measuring cylinders, tall form, class A, borosilicate glass
Cylinders Measuring Cylinders
DURAN®, borosilicate glass, with PE stopper, class A.
  • Calibrated to contain 'ln', certificate of conformity (DE-M marking) with dated batch identification available via website: (www.hirschmannlab.de)
  • Hexagonal glass base, NS ground joint
  • Blue graduations with ring marks at major graduations
DIN EN ISO 4788
Certifications: DIN EN ISO 4788
Order Now
SPECIFICATIONS
Environmentally Preferable