Dimethyl sulphoxide ≥99.5% stationary phase for GC

Supplier: VWR Chemicals

Synonyms: Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO

23487.291EA 242 EUR
23487.291
Dimethyl sulphoxide ≥99.5% stationary phase for GC
Dimethyl sulphoxide
Formula: (CH₃)₂SO
MW: 78.14 g/mol
Boiling Pt: 189
Melting Pt: 18.5 °C
Density: 1.101 g/cm³ (20 °C)
Flash Pt: 87
Storage Temperature: Ambient
MDL Number: MFCD00002089
CAS Number: 67-68-5
EINECS: 200-664-3
REACH: 01-2119431362-50
Merck Index: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR