1,4-Dioxane, dehydrated (max. 0.01% H₂O) ≥99.5% stabilised, AnalaR NORMAPUR® analytical reagent

Supplier: VWR Chemicals
Danger

Synonyms: Glycolethylether, 1,4-Dioxacyclohexane, Dioxane, p-Dioxane, 1,4-Diethylene dioxide

Stabilisation: 20 ppm Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

23539.291EA 166 EUR
23539.291
1,4-Dioxane, dehydrated (max. 0.01% H₂O) ≥99.5% stabilised, AnalaR NORMAPUR® analytical reagent
1,4-Dioxane
Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 101.1 °C (1013 hPa)
Melting Pt: 11.8 °C
Density: 1.03 g/cm³ (20 °C)
Flash Pt: 12
Storage Temperature: Ambient
MDL Number: MFCD00006571
CAS Number: 123-91-1
EINECS: 204-661-8
UN: 1165
ADR: 3 II
Merck Index: 13,03330
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df424642d4a7a96d2eebfb99cf9a2e6d128185d0&vid=37c3b8119f755f99624576b4e975000cbbb8349d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99.5 %
Acidity or alkalinity Max. 0.001 meq/g
Colouration Max. 10 APHA
Peroxides (as H2O2) Max. 0.0100 %
Water Max. 0.0100 %
Acetyls and carbonyl derivatives Max. 40 ppm
Evaporation residue Max. 50 ppm
Fe (Iron) Max. 1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR