Hydrogen peroxide 50% stabilised, GPR RECTAPUR®

Supplier: VWR Chemicals
Danger

23620.292EA 94.5 EUR
23620.292
Hydrogen peroxide 50% stabilised, GPR RECTAPUR®
Hydrogen peroxide
Formula: H₂O₂
MW: 34.01 g/mol
Boiling Pt: 150.2
Melting Pt: –40 °C
Density: 1.19 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011333
CAS Number: 7722-84-1
EINECS: 231-765-0
UN: 2014
ADR: 5.1 (8) II
Merck Index: 14,04798

Order Now

Specification Test Results

Assay 48 - 51 %
Colouration Max. 10 APHA
Evaporation residue Max. 0.05 %
Heavy metals (as Pb) Max. 5 ppm
Total N (Nitrogen) Max. 40 ppm
Cl (Chloride) Max. 5 ppm
Ca (Calcium) Max. 5 ppm
Fe (Iron) Max. 5 ppm
Mg (Magnesium) Max. 5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR