Aluminium chloride hexahydrate 99%, Reagent Grade

Supplier: Honeywell Chemicals
Warning

Synonyms: Aluminum chloride hexahydrate

237078-100G 237078-500G
237078-100GEA 26.1 EUR
237078-100G 237078-500G
Aluminium chloride hexahydrate 99%, Reagent Grade
Aluminium chloride hexahydrate

  • ≥95% for general laboratory use.

Formula: AlCl₃·6H₂O
Melting Pt: ∼100 °C
Density: 2,4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149134
CAS Number: 7784-13-6
EINECS: 231-208-1
ADR: 8,III

Order Now

Specification Test Results

Vapor Pressure 1 mmHg ( 100 °C)
Assay 98.5 - 101.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR