Petroleum spirit, 40…60 °C, AnalaR NORMAPUR® ACS analytical reagent (max. 0.01% aromatic hydrocarbons)

Supplier: VWR Chemicals
Danger

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

23835.294EA 58.8 EUR
23835.294 23835.328 VWRC23835.363_D 23835.460 23835.556 23835.363
Petroleum spirit, 40…60 °C, AnalaR NORMAPUR® ACS analytical reagent (max. 0.01% aromatic hydrocarbons)
Petroleum spirit
Boiling Pt: 40…60 °C (1013 hPa)
Density: 0.645…0.665 g/cm³ (20 °C)
Flash Pt: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
EINECS: 265-151-9
UN: 1268
ADR: 3 II
REACH: 01-2119474679-18

Order Now

Specification Test Results

Acidity Max. 0.0002 meq/g
Bromine value Max. 1
Colouration Max. 10 APHA
Boiling range (40-60°C) (Vol %) Min. 90%
Substances coloured by H2SO4 Max. 150 APHA
Benzene Max. 100 ppm
Evaporation residue Max. 10 ppm
Total S (as SO4) Max. 50 ppm
Water Max. 100 ppm
Al (Aluminium) Max. 0.2 ppm
Ba (Barium) Max. 0.1 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.05 ppm
Co (Cobalt) Max. 0.02 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.02 ppm
Fe (Iron) Max. 0.1 ppm
K (Potassium) Max. 0.3 ppm
Mg (Magnesium) Max. 0.1 ppm
Mn (Manganese) Max. 0.02 ppm
Na (Sodium) Max. 1 ppm
Ni (Nickel) Max. 0.02 ppm
Pb (Lead) Max. 0.1 ppm
Sn (Tin) Max. 0.1 ppm
Sr (Strontium) Max. 0.03 ppm
Zn (Zinc) Max. 0.1 ppm
Conforms to ACS Passes test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR