Burette clamps, Stronghold™
Supplier: HARVARD APPARATUS
53-2170-W
241-2511EA 144 EUR
241-2511
Burette clamps, Stronghold™
Clamps General Purpose Clamps
These burette clamps are made from precision injection molded nylon with 30% glass fiber reinforcement which provides enormous strength at a great saving in weight.
  • Material is perfect for laboratory use since it has the very high melting point of 221 °C
  • Good resistance to chemicals, particularly salt solutions and most solvents
  • Clamps with red body feature white thumb screws, clamps with white body feature red thumb screws
  • Corrosion resistant
The single burette clamp has two small clamps mounted vertically 100 mm apart to hold firmly in two placesMore Product Information