Sodium acetate ≥99.0% ACS, Fluka™

Supplier: Honeywell Chemicals

241245-1KG 241245-2.5KG
241245-1KGEA 272.56 EUR
241245-1KG 241245-2.5KG
Sodium acetate ≥99.0% ACS, Fluka™
Sodium acetate
Formula: H₃CCOONa
Boiling Pt: Dec. (on heating)
Melting Pt: 324 °C
Density: 1,45 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012459
CAS Number: 127-09-3
EINECS: 204-823-8
REACH: 01-2119485123-42
Merck Index: 12,08711

Order Now

Specification Test Results

Assay (HClO₄-titr.) Min. 99.0 %
Water insoluble matter Max. 0.01 %
Loss on drying (120°C) Max. 1.0 %
pH (5 %, 25 °C) 7.0 - 9.2
Calcium (Ca) Max. 0.005 %
Iron (Fe) Max. 0.001 %
Magnesium (Mg) Max. 0.002 %
Heavy metals (as Pb) Max. 0.001 %
Chloride (Cl) Max. 0.002 %
Phosphate (PO₄) Max. 0.001 %
Sulfate (SO₄) Max. 0.003 %
IR spectrum complying
Form Solid
pKa  4.76 (acetic acid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR