Iron(II) sulphate heptahydrate 96, TECHNICAL

Supplier: VWR Chemicals
Warning

Synonyms: Iron vitriol, Ferrous sulphate heptahydrate

24237.294EA 39.3 EUR
24237.294 24237.363
Iron(II) sulphate heptahydrate 96, TECHNICAL
Iron (II) sulphate heptahydrate
Formula: FeSO₄·7H₂O
MW: 278.02 g/mol
Melting Pt: 64
Density: 1.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149719
CAS Number: 7782-63-0
EINECS: 231-753-5
Merck Index: 13,04091

Order Now

Specification Test Results

Assay Min. 96 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR