Glycerine 98, GPR RECTAPUR®

Supplier: VWR Chemicals

Synonyms: 1,2,3-Propanetriol, Propane-1,2,3-triol, Glycerol

24387.292EA 79.6 EUR
24387.292 24387.326 24387.361 24387.463 24387.554
Glycerine 98, GPR RECTAPUR®
Glycerine
Formula: HOCH₂CH(OH)CH₂OH
MW: 92.09 g/mol
Boiling Pt: 290
Melting Pt: 18.6 °C
Density: 1.26 g/cm³ (20 °C)
Flash Pt: 177 °C (open cup)
Storage Temperature: Ambient
MDL Number: MFCD00004722
CAS Number: 56-81-5
EINECS: 200-289-5
Merck Index: 13,04497
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=86a15453b54e7106c121d32e4e716385003c4507&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 98 %
Density (20/4) 1.257 - 1.265
Heavy metals (as Pb) Max. 20 ppm
Fe (Iron) Max. 10 ppm
Ignition residue (SO4) Max. 0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR