FAM Benzine, 65-95 °C DIN 51635, Riedel-de Haën™

Supplier: Honeywell Chemicals
Danger

Synonyms: FAM-Petroleum ether, Petroleum benzin, Petroleum ether, Petroleum spirit

24538-18L 24538-2.5L 24538-25L 24538-4X2.5L
24538-18LEA 1430 EUR
24538-18L 24538-2.5L 24538-25L 24538-4X2.5L
FAM Benzine, 65-95 °C DIN 51635, Riedel-de Haën™
FAM benzine

UNSPSC Code: 12191502

Boiling Pt: 65…95 °C
Melting Pt: –30 °C
Density: 0.76 g/cm³ (15 °C)
Flash Pt: –26 °C
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
UN: 3295
ADR: 3 II

Order Now

Specification Test Results

Boiling range 60 - 95 °C
Density (15 °C) 0.695 - 0.710 g/cm³
Amount of distillate 90 °C 85 - 95 Vol.%
Residue from evaporation (DIN EN ISO 6246) Max. 0.03 g/l
n-Hexane (GC) Max. 5 %
Benzene (GC) Max. 0.1 %
APHA Max. 10


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR