Oil 10 S, VOLTALEF®

Supplier: VWR Chemicals

24627.188EA 540 EUR
24627.188
Oil 10 S, VOLTALEF®
Poly(chlorotrifluoroethylene) 800
Formula: (C₂ClF₃)n
MW: 800 g/mol
Density: 1.9 g/cm³ (20 °C)
CAS Number: 9002-83-9

Order Now

Specification Test Results

Identification Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR