Hydroxylammonium chloride, TECHNICAL

Supplier: VWR Chemicals
Warning

Synonyms: Hydroxylamine HCl, Hydroxylamine hydrochloride

24706.238EA 65.9 EUR
24706.238
Hydroxylammonium chloride, TECHNICAL
Hydroxylammonium chloride
Formula: NH₂OH·HCl
MW: 69.49 g/mol
Boiling Pt: 306
Melting Pt: 151
Density: 1.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00051089
CAS Number: 5470-11-1
EINECS: 226-798-2
UN: 2923
ADR: 8 (6.1) III
Merck Index: 13,04853

Order Now

Specification Test Results

Identification Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR