N-(1-Naphthyl)ethylenediamine dihydrochloride 98,0 zA

Supplier: VWR Chemicals
Warning

Synonyms: Marshall's Reagent, N-2-Aminoethyl-1-naphthylamine dihydrochloride, N-1-Naphthylethylenediamine

25792.130EA 221 EUR
25792.130 25792.120
N-(1-Naphthyl)ethylenediamine dihydrochloride 98,0 zA
N-2-Aminoethyl-1-naphthylamine dihydrochloride
Formula: C₁₂H₁₆Cl₂N₂
MW: 259.18 g/mol
Melting Pt: 188…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00012556
CAS Number: 1465-25-4
EINECS: 215-981-2
Merck Index: 12,06496
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3643e6df3e9cf8dc627823e9090c97004ee25308&vid=a30fc28279c11af12c6b735b00b84fc8f08caf34&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (calculated on dried substance) Min. 98.0 %
IR Spectrum Passes test
Sensitivity to sulphanilamide Passes test
Solubility Passes test
Loss on drying (80°C; 4 h) Max. 5 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR