Ninhydrin, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. zA

Supplier: VWR Chemicals
Warning

Synonyms: Ninhydrin monohydrate, Trioxohydrindene monohydrate, 2,2-Dihydroxyindane-1,3-dione, 2,2-Dihydroxy-1,3-indanedione, Indanetrione hydrate, 2,2-Dihydroxyindan-1,3-dion, 1,2,3-Indantrione monohydrate

25905.107EA 29.3 EUR
25905.107 25905.153 25905.180
Ninhydrin, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. zA
Ninhydrin
Formula: C₉H₆O₄
MW: 178.14 g/mol
Boiling Pt: 351
Melting Pt: 250…258 °C
Density: 0.862 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003791
CAS Number: 485-47-2
EINECS: 207-618-1
Merck Index: 13,06582
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0ff1a5cd0c772262dac61a6c63dbdeca97baa2c&vid=61d6c7dbd1fe019587f026a878ce0951ce80fcc8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay min 99%
Identification and melting point Passes test
IR Spectrum Passes test
Solubilty in water (10 g/l) Passes test
Suited for amino acid reagent Passes test
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR