n-Pentane 99,0, AnalaR NORMAPUR® zA

Supplier: VWR Chemicals
Danger

Synonyms: Pentane

26185.297EA 113 EUR
26185.297 26185.322 26185.460
n-Pentane 99,0, AnalaR NORMAPUR® zA
n-Pentane
Formula: H₃C(CH₂)₃CH₃
MW: 72.15 g/mol
Boiling Pt: 35…36 °C (1013 hPa)
Melting Pt: –130 °C
Density: 0.63 g/cm³ (20 °C)
Flash Pt: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00009498
CAS Number: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3 II
REACH: 01-2119459286-30
Merck Index: 13,07193
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ba2c9c81a4b1f44769627a00ba49b6cc8e63132&vid=f6831eb15ab1602d8c752468dc3ee31cc4ee61c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (on anhydrous substance) Min. 99.0 %
Assay (C5 isomers) Min. 99.9 %
Acidity Max. 0.004 meq/g
Boiling point 35 - 37 °C
Colouration Max. 10 APHA
Density (20/4) 0.625 - 0.632
Aromatics (as C6H6) Max. 50 ppm
Evaporation residue Max. 10 ppm
Total S (as SO4) Max. 50 ppm
Water Max. 100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR