Pumice stone, granules zA, washed

Supplier: VWR Chemicals

26398.293EA 101 EUR
26398.293 26398.362
Pumice stone, granules zA, washed
Pumice stone
Melting Pt: > 500 °C
MDL Number: MFCD00148147
CAS Number: 1332-09-8

Order Now

Specification Test Results

Heavy metals (as Pb) Max. 50 ppm
Solubility in water Max. 0.5 %
Soluble chlorides Max. 0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR