Volumetric flasks, class A, USP, white markings
Supplier: HIRSCHMANN
264016145 264016545 264017045 264017545 264018045 264018545 264015327 264016127 264016527 264017027 264017527 264018027 264018527 264018627 264018927 264019327 264019427
264016145.EA 63.9 EUR
264016145. 264016545. 264017045. 264017545. 264018045. 264018545. HIRS264015327 HIRS264016127 HIRS264016527 HIRS264017027 HIRS264017527 HIRS264018027 HIRS264018527 HIRS264018627 HIRS264018927 HIRS264019327 HIRS264019427
Volumetric flasks, class A, USP, white markings
Flasks Volumetric Flasks
DURAN®, borosilicate glass, amber, with PE stopper, USP certified.
  • White enamelled indelible markings resistant to acids and alkalis
  • USP, with dated batch identification
  • Additional USP batch certificates can be generated and printed at any time free of charge at www.hirschmannlab.de
Certifications: DIN EN ISO 1042
Order Now
SPECIFICATIONS
Environmentally Preferable