Peqlab peqGOLD, Protein Marker V (Pre-Stained)
Supplier: VWR Chemicals
27-2210EA 185 EUR
27-2210
Peqlab peqGOLD, Protein Marker V (Pre-Stained)
Electrophoresis Reagents Molecular Weight Markers
Protein Marker V (pre-stained) for monitoring protein separation, transfer efficiency and size determination.
  • Mix of nine recombinant proteins
  • Covalently pre-stained by using one of three different dyes
  • Range: 10 to 250 kDa
  • Concentration: 100 to 200 ng/µl each protein
  • Ready to use for a fast, convenient workflow