Adapters, receiver, with side socket, Quickfit®
Supplier: SciLabware
RA4/11
271-0012EA 99.1 EUR
271-0012
Adapters, receiver, with side socket, Quickfit®
Distillation Components Distillation Adapters
PYREX®, Borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With side socket inclined at an angle of 105°.