Sodium acetate trihydrate ≥98%, GPR RECTAPUR®

Supplier: VWR Chemicals

27648.294EA 51.2 EUR
27648.294 27648.363
Sodium acetate trihydrate ≥98%, GPR RECTAPUR®
Sodium acetate trihydrate
Formula: H₃CCOONa·3H₂O
Boiling Pt: 120…123 °C (1013 hPa)
Melting Pt: 58 °C
Density: 1,4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00071557
CAS Number: 6131-90-4
EINECS: 204-823-8
Merck Index: 13,08642
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=313c1d45673fab192565649f8cbb7ddff555de39&vid=1ad37a966605626c95117735c83b16eaea381fbc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 98 %
Heavy metals (as Pb) Max. 20 ppm
Cl (Chloride) Max. 0.02 %
SO4 (Sulphate) Max. 100 ppm
Fe (Iron) Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR