Sodium hydrogen carbonate 98, TECHNICAL

Supplier: VWR Chemicals

Synonyms: Sodium bicarbonate

27780.291EA 23.3 EUR
27780.291 27780.360
Sodium hydrogen carbonate 98, TECHNICAL
Sodium hydrogen carbonate
Formula: NaHCO₃
MW: 84.01 g/mol
Melting Pt: 270
Density: 2.159 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003528
CAS Number: 144-55-8
EINECS: 205-633-8
Merck Index: 13,08655
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 98 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR