Protein A Mag Sepharose
Supplier: Cytiva
28-9513-78 28-9440-06
28-9513-78EA 154 EUR
28-9513-78 28-9440-06
Protein A Mag Sepharose
Beads Magnetic Beads
Protein A Mag Sepharose magnetic beads are designed to simplifying enrichment of target proteins via immunoprecipitation or pull-down assays used to detect physical interactions between two or more proteins.
  • Visible and dense Sepharose based magnetic beads with protein A as the ligand make it easy to spot and collect bound target protein
  • Non-adherent beads eliminate smearing effects and aggregate formation
  • Can be used without detergents
  • Simple capture of target protein in small or large sample volumes (low microliter to high milliliter scale)
  • Optimized
More Product Information