Sodium tetraphenylborate 99,5 zA

Supplier: VWR Chemicals
Danger

Synonyms: Tetraphenyl sodium borate, Sodium tetraphenylboron, Tetraphenylboron sodium salt

28187.138EA 230 EUR
28187.138
Sodium tetraphenylborate 99,5 zA
Sodium tetraphenylborate
Formula: (C₆H₅)₄BNa
MW: 342.22 g/mol
Melting Pt: > 400 °C
Density: 1.15 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011494
CAS Number: 143-66-8
EINECS: 205-605-5
UN: 2811
ADR: 6.1 III
Merck Index: 13,08765

Order Now

Specification Test Results

Assay Min. 99.5 %
Loss on drying (100°C) Max. 0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR