Volumetric flasks, class A, blue markings
Supplier: HIRSCHMANN
2820166 2820197 2820198 2820152 2820153 2820160 2820161 2820165 2820170 2820171 2820175 2820178 2820180 2820181 2820185 2820186 2820190 2820192 2820193 2820194 2820182 282015208 282016008 282016108 282016508 282017008 282017508 282017808 282018008 282018108 282018508 282018608 282019008 282019308 282019408 282019445 282019708 282019808
2820166.EA 20.4 EUR
2820166. 2820197. 2820198. 2820152. 2820153. 2820160. 2820161. 2820165. 2820170. 2820171. 2820175. 2820178. 2820180. 2820181. 2820185. 2820186. 2820190. 2820192. 2820193. 2820194. 2820182. 282015208. 282016008. 282016108. 282016508. 282017008. 282017508. 282017808. 282018008. 282018108. 282018508. 282018608. 282019008. 282019308. 282019408. 282019445. 282019708. 282019808.
Volumetric flasks, class A, blue markings
Flasks Volumetric Flasks
DURAN®, borosilicate glass, with PE stopper, conformity certified (DE-M marking).
  • Blue enamel indelible markings resistant to acids and alkalis
  • Certificate of conformity with dated batch identification available via website (www.hirschmannlab.de)
* In addition to DIN.
**Wide neck
Certifications: DIN EN ISO 1042
Order Now
SPECIFICATIONS
Environmentally Preferable